Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PHNPALAAAA LDYRCPVHFY WVRGEEIIPR GHRRGRIDDL RYQIDDKPNN QIRISKQLAE FVPLDYSVPI EIPTIKCKPD KLPLFKRQYE NHIFVGSKTA DPCCYGHTQF HLLPDKLRRE RLLRQNCADQ IEVVFRANAI ASLFAWTGAQ AMYQGFWSEA DVTRPFVSQA VITDGKYFSF FCYQLNTLAL TTQADQNNPR KNICWGTQSK PLYETIEDND VKGFNDDVLL QIVHFLLNRP KEEKSQLLEN; Positive Samples: SW620, HL-60, HeLa, HepG2; Cellular Location: Mitochondrion
Mitochondrial ribosomes consist of a large 39S subunit and a small 28S subunit, both of which are comprised of multiple mitochondrial ribosomal proteins (MRPs) that are encoded by nuclear genes and are essential for protein synthesis within mitochondria. MRP-S30 (mitochondrial ribosomal protein S30), also known as PDCD9 (programmed cell death protein 9), is a 439 amino acid protein that localizes to the mitochondrion, where it exists as a component of the 28S ribosomal subunit and works in conjunction with other MRPs to mediate protein synthesis. MRP-S30 is expressed in kidney, liver, heart and skeletal muscle. The gene encoding MRP-S30 maps to human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28S ribosomal protein S30, mitochondrial; 39S ribosomal protein S30, mitochondrial; Large ribosomal subunit protein mL65; Large ribosomal subunit protein mS30; MRP S30; MRP-S30; programmed cell death 9; Programmed cell death protein 9; S30mt
Gene Aliases: 2610020A16Rik; AA968347; BM-047; MRP-S30; MRPS30; PAP; PDCD9; S30mt
UniProt ID: (Human) Q9NP92, (Mouse) Q9D0G0
Entrez Gene ID: (Human) 10884, (Mouse) 59054, (Rat) 294767
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support