Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MECLRSLPCL LPRAMRLPRR TLCALALDVT SVGPPVAACG RRANLIGRSR AAQLCGPDRL RVAGEVHRFR TSDVSQATLA SVAPVFTVTK FDKQGNVTSF VFESCDNSRV SSDIRLS
Mrs2 is a magnesium transporter that mediates the influx of magnesium into the mitochondrial matrix. Mrs2 is required for normal expression of the mitochondrial respiratory complex I subunits, and mediates the influx of magnesium into the mitochondrial matrix. Mrs2 is also required for normal expression of the mitochondrial respiratory complex I subunits.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Magnesium transporter MRS2 homolog, mitochondrial; MRS2 magnesium transporter; MRS2-like protein; MRS2-like, magnesium homeostasis factor; putative magnesium transporter
Gene Aliases: HPT; MRS2; MRS2L
UniProt ID: (Human) Q9HD23
Entrez Gene ID: (Human) 57380
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support