Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
May be involved in signal transduction as a component of a multimeric receptor complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD20 antigen-like 3; CD20-like precusor; Four-span transmembrane protein 3; four-span transmembrane protein 3.1; four-span transmembrane protein 3.2; HAIRB-iso; Membrane-spanning 4-domains subfamily A member 6A; membrane-spanning 4-domains subfamily A member 6D; membrane-spanning 4-domains, subfamily A, member 11; membrane-spanning 4-domains, subfamily A, member 6A; membrane-spanning 4-domains, subfamily A, member 6A, isoform 2; MS4A6A-polymorph
Gene Aliases: 4SPAN3; 4SPAN3.2; CD20L3; CDA01; Ms4a11; MS4A6; MS4A6A; MST090; MSTP090
UniProt ID: (Human) Q9H2W1
Entrez Gene ID: (Human) 64231, (Rat) 361735
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support