Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • MSX1/MSX2 Antibodies

          Invitrogen

          MSX1/MSX2 Polyclonal Antibody

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MSX1/MSX2 Polyclonal Antibody

          • Antibody Testing Data (2)
          MSX1/MSX2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MSX1/MSX2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MSX1/MSX2 Antibody (PA5-103302) in ICC/IF

          Immunocytochemistry analysis of MSX1/MSX2 in A549 cells. The samples were fixed with PFA and permeabilized in 0.1% Triton X-100, then blocked in 10% serum for 45 minutes at 25°C. Samples were incubated with MSX1/MSX2 polyclonal antibody (Product # PA5-103302) and mouse anti-beta tubulin antibody for 1 hour at 37°C. An AlexaFluor594 conjugated goat anti-rabbit IgG(H+L) (Red) and an AlexaFluor488 conjugated g... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MSX1/MSX2 Antibody in Immunocytochemistry (ICC/IF)
          MSX1/MSX2 Antibody in Western Blot (WB)

          Product Details

          PA5-103302

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:3,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human MSX1(Accession P28360), corresponding to amino acid residues W145-Q195.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          RRID

          AB_2852649

          Product Specific Information

          Antibody detects endogenous levels of total MSX1/2.

          Target Information

          Drosophila,muscle segment (msh) homolog 1,homeo domain encoding gene,inhibiting MYOD1 expression,highly expressed in dental mesenchyme during critical bud stage,involved in epithelial-mesenchymal signaling in many organs,and in the pathogenesis of cleft lip and palate,interacting with MSX2 in mouse limb bud patterning .This gene encodes a member of the muscle segment homeobox gene family. The encoded protein functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins. It may also have roles in limb-pattern formation, craniofacial development, particularly odontogenesis, and tumor growth inhibition.Tissue specificity: Expressed in the developing nail bed mesenchyme.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: homeo box, msh-like 1; homeo box, msh-like 2; homeobox 7; Homeobox protein Hox-7; Homeobox protein Hox-8; Homeobox protein Hox-8-1; Homeobox protein MSX-1; Homeobox protein MSX-2; homeobox, msh-like 1; homeobox, msh-like 2; Hox-7.1; hox-8.1; msh homeo box 1; msh homeo box 2; msh homeo box homolog 2; Msh homeobox 1-like protein; msh homeobox homolog 1; msh homeobox homolog 2; muscle-segment homeobox

          View more View less

          Gene Aliases: AA675338; AI324650; BB122635; CRS2; ECTD3; FPP; Hox-7; Hox-8; Hox-8.1; HOX7; Hox7.1; HOX8; Hox8.1; HYD1; MSH; Msx-2; MSX1; MSX2; PFM; PFM1; STHAG1

          View more View less

          UniProt ID: (Human) P28360, (Human) P35548, (Mouse) P13297, (Mouse) Q03358

          View more View less

          Entrez Gene ID: (Human) 4487, (Human) 4488, (Mouse) 17701, (Rat) 81710, (Mouse) 17702, (Rat) 25483

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding p53 binding RNA polymerase II core promoter proximal region sequence-specific DNA binding transcription cofactor activity protein binding transcription factor binding sequence-specific DNA binding transcription regulatory region DNA binding DNA binding molecular_function transcription factor activity, transcription factor binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter cell morphogenesis in utero embryonic development epithelial to mesenchymal transition involved in endocardial cushion formation transcription from RNA polymerase II promoter muscle organ development negative regulation of cell proliferation anterior/posterior pattern specification mesenchymal cell proliferation signal transduction involved in regulation of gene expression negative regulation of cell growth positive regulation of BMP signaling pathway forebrain development midbrain development protein localization to nucleus embryonic forelimb morphogenesis embryonic hindlimb morphogenesis embryonic nail plate morphogenesis middle ear morphogenesis odontogenesis of dentin-containing tooth regulation of odontogenesis negative regulation of apoptotic process positive regulation of DNA damage response, signal transduction by p53 class mediator positive regulation of transcription from RNA polymerase II promoter stem cell differentiation protein stabilization negative regulation of striated muscle cell differentiation palate development face morphogenesis bone morphogenesis cartilage morphogenesis mammary gland epithelium development BMP signaling pathway involved in heart development activation of meiosis positive regulation of intrinsic apoptotic signaling pathway by p53 class mediator negative regulation of transcription regulatory region DNA binding positive regulation of mesenchymal cell apoptotic process osteoblast differentiation chondrocyte development osteoblast development outflow tract septum morphogenesis endochondral bone growth negative regulation of CREB transcription factor activity wound healing, spreading of epidermal cells negative regulation of fat cell differentiation negative regulation of keratinocyte differentiation positive regulation of osteoblast differentiation negative regulation of transcription, DNA-templated positive regulation of catagen bone trabecula formation cranial suture morphogenesis frontal suture morphogenesis branching involved in mammary gland duct morphogenesis enamel mineralization cellular response to estradiol stimulus ossification outflow tract morphogenesis transcription, DNA-templated regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter multicellular organism development embryonic limb morphogenesis BMP signaling pathway wound healing odontogenesis anagen embryonic digit morphogenesis regulation of apoptotic process positive regulation of apoptotic process cartilage development cellular response to growth factor stimulus epithelial to mesenchymal transition heart morphogenesis heart development negative regulation of DNA binding pituitary gland development cellular response to nicotine
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0