Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: DRGCGFPVGE HSPHSRVQIG DHSLRLQTAD RGQPHKQVVE NQQLFSAFKA LLEDFRAELR EDERARLRLQ QQYASDKAAW DVEWAVLKCR LEQ
Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 80%.
Microtubule-associated factor involved in the late phase of epithelial polarization and microtubule dynamics regulation. Plays a role in the development and maintenance of non-centrosomal microtubule bundles at the lateral membrane in polarized epithelial cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: coiled-coil domain containing 165; Coiled-coil domain-containing protein 165; Microtubule cross-linking factor 1; PAR-1-interacting protein; protein SOGA2; SOGA family member 2
Gene Aliases: CCDC165; KIAA0802; MTCL1; SOGA2
UniProt ID: (Human) Q9Y4B5
Entrez Gene ID: (Human) 23255
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support