Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Cow: 86%; Dog: 79%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%
Myeloid tumor related gene (MTGR1) is found deleted in acute myeloid leukemia. Overexpression of MTGR1 can stimulate AML1-MTG8 to induce GCSF- dependent proliferation of L-G cells and to interfere with AML1- dependent transcription. AML1/MTG8 functions as a complex with MTGR1 and that the complex might be important in promoting leukemogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CBFA2T2 identified gene homolog; core-binding factor runt domain alpha subunit 2 translocated to 2 homolog; core-binding factor, runt domain, alpha subunit 2, translocated to, 2 homolog; DKFZp313F2116; MTG8-like protein; MTG8-related protein 1; Protein CBFA2T2; RP5-1137F22.1; ZMYND3 CBFA2T2
Gene Aliases: A430091M07; C330013D05Rik; Cbfa2t2; Cbfa2t2h; MTGR1
UniProt ID: (Mouse) O70374
Entrez Gene ID: (Mouse) 12396
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support