Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LSIPYPGCEF FKEYKDRDYM AEGLIFNWKQ DYVDAPLSIP DFLTHSLNID WSQYQCWDLV QQTQNYLKLL LSLVNSDDDS GLLVHCISGW DRTPLFISLL RLSLWADGLI HTSLKPTEIL YLTVAYDWFL FGHMLVDRLS KGEEIFFFCF NFLKHITSEE FSALKTQRRK SLPARDGGFT LEDICMLRRK DRGSTTSLGS DFSLVMESSP GATGSFTYEA VELVPAGAPT QAAWRKSHSS SPQSVLWNRP QPSEDRLPSQ QGLAEARSSS SSSSNHSDNF FRMGSSPLEV PKPRLAALSD RETRLQEVRS AFLAAYSSTV GLRAVAPSPS GAIGGLLEQF ARGVGLRSIS SNAL
This gene encodes a myotubularin-related protein. The encoded protein is a phosphoinositide phosphatase that specifically dephosphorylates phosphatidylinositol 3,5-biphosphate and phosphatidylinositol 3-phosphate. Mutations in this gene are correlated with autosomal dominant centronuclear myopathy. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 18.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: egg-derived tyrosine phosphatase homolog; HCV NS5A-transactivated protein 4 splice variant A-binding protein 1; hJumpy; jumpy; mJumpy; Myotubularin-related protein 14; NS5ATP4ABP1; Phosphatidylinositol-3,5-bisphosphate 3-phosphatase MTMR14; Phosphatidylinositol-3-phosphate phosphatase
Gene Aliases: 1110061O04Rik; AW553738; C3orf29; C76151; MTMR14; RGD1304842
UniProt ID: (Human) Q8NCE2, (Mouse) Q8VEL2
Entrez Gene ID: (Human) 64419, (Mouse) 97287, (Rat) 312634
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support