Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MQLQEQFDNL NAAHETSKLE IEASHSEKLE LLTKAYEASL SEIKKGHEIE KKSLEDLLSE KQESLEKQIN DLKSENDALN EKLKSEEQKR RAREKANLKN PQIMYLEQEL ESLKAVLEIK NEKLHQQDIK LMKMGKLVDN NTALVDKLKR FQQENEELKA RMDKHMAISR QLSTEQAVLQ ESLEKESKVN KRLSMENEEL LWKLHNGDLC SPKRSPTSSA IPLQSPRNSG SFPSPSISPR
The protein belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase family. It is located in endoplasmic reticulum and acts as molecular chaperones.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Angiotensin-II type 2 receptor-interacting protein; AT2 receptor-binding protein; AT2 receptor-interacting protein; AT2R binding protein; DKFZp586D1519; DKFZp686F20243; erythroid differentiation-related; FLJ14295; Microtubule-associated tumor suppressor 1; Mitochondrial tumor suppressor 1; mitochondrial tumor suppressor gene 1; transcription factor MTSG1
Gene Aliases: ATBP; ATIP; GK1; ICIS; KIAA1288; MP44; MTSG1; MTUS1
UniProt ID: (Human) Q9ULD2
Entrez Gene ID: (Human) 57509
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support