Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Mucin 13, epithelial transmembrane. SEA is an extracellular domain associated with O-glycosylation. Proteins found to contain SEA-modules include, agrin, enterokinase, 63 kDa sea urchin (Strongylocentrotus purpuratus) sperm protein, perlecan (heparan sulphate proteoglycan core, mucin 1 and the cell surface antigen, 114/A10, and two functionally uncharacterised, probably extracellular, Caenorhabditis elegans proteins. Despite the functional diversity of these adhesive proteins, a common denominator seems to be their existence in heavily glycosylated environments. In addition, the better characterised proteins all contain O-glycosidic-linked carbohydrates such as heparan sulphate that contribute considerably to their molecular masses. The common module might regulate or assist binding to neighbouring carbohydrate moieties. Enterokinase, the initiator of intestinal digestion, is a mosaic protease composed of a distinctive assortment of domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell surface antigen 114/A10; Down-regulated in colon cancer 1; Lymphocyte antigen 64; MUC-13; mucin 13; mucin 13, epithelial transmembrane; Mucin-13
Gene Aliases: 114/A10; 14/A10; AI159736; DRCC1; Lrrp; Ly64; Muc-123; MUC-13; MUC13; NJ-1; RECC; UNQ6194/PRO20221
UniProt ID: (Human) Q9H3R2, (Mouse) P19467
Entrez Gene ID: (Human) 56667, (Rat) 207126, (Mouse) 17063
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support