Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: SVYRQKARV SQELKGAKKV HLGEDLKSIL SEAPGKCVPY AVIEGAVRSV KETLNSQFVE NCKGVIQRLT LQEHKMVWNR TTHLWNDCSK IIHQRTNTVP FDLVPHEDGV DVAVRVLKPL DSVDLGLETV YEKFHPSIQS FTDVIGHYIS GERPKGIQET EEMLKVGATL TGVGELVLDN NSVRLQPPKQ GMQYYLSSQD FDSLLQRQES SVRL (27-239 aa encoded by BC010101)
E3 ubiquitin-protein ligase that plays a role in the control of mitochondrial morphology. Promotes mitochondrial fragmentation and influences mitochondrial localization. Inhibits cell growth. When overexpressed, activates JNK through MAP3K7/TAK1 and induces caspase-dependent apoptosis. E3 ubiquitin ligases accept ubiquitin from an E2 ubiquitin-conjugating enzyme in the form of a thioester and then directly transfer the ubiquitin to targeted substrates.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 SUMO-protein ligase MUL1; E3 ubiquitin ligase; E3 ubiquitin-protein ligase MUL1; Growth inhibition and death E3 ligase; mitochondria-anchored protein ligase; mitochondrial E3 ubiquitin ligase 1; mitochondrial ubiquitin ligase activator of NF-kB; Mitochondrial ubiquitin ligase activator of NFKB 1; Mitochondrial-anchored protein ligase; Protein Hades; Putative NF-kappa-B-activating protein 266; RING finger protein 218; RING-type E3 ubiquitin transferase NFKB 1; RP11-401M16.2
Gene Aliases: 0610009K11Rik; AV000801; C1orf166; GIDE; MAPL; MUL1; MULAN; RGD1309944; RNF218
UniProt ID: (Human) Q969V5, (Mouse) Q8VCM5
Entrez Gene ID: (Human) 79594, (Mouse) 68350, (Rat) 298576
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support