Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAAAVRMNI QMLLEAADYL ERREREAEHG YASMLPYNNK DRDALKRRNK SKKNNSSSRS THNEMEKNRR AHLRLCLEKL KGLVPLGPES SRHTTLSLLT KAKLHIKKLE DCDRKAVHQI DQLQREQRHL KRQLEKLGIE RIRMDSIGST VSSERSDSDR EEIDVDVEST DYLTGDLDWS SSSVSDSDER GSMQSLGSDE GYSSTSIKRI KLQDSHKACL GL (1-221 aa encoded by BC098396)
MAX dimerization protein belongs to a subfamily of MAX-interacting proteins. This protein competes with MYC for binding to MAX to form a sequence-specific DNA-binding complex, acts as a transcriptional repressor (while MYC appears to function as an activator) and is a candidate tumor suppressor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antagonizer of myc transcriptional activity; Max dimerization protein 1; Max dimerizer 1; MAX-binding protein; MGC104659; OTTHUMP00000202211; Protein MAD
Gene Aliases: AW122478; BHLHC58; MAD; Mad-1; MAD1; MXD1
UniProt ID: (Human) Q05195
Entrez Gene ID: (Human) 4084, (Rat) 362391, (Mouse) 17119
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support