Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • MYBBP1A Antibodies

          Abnova

          MYBBP1A Polyclonal Antibody, MaxPab™

          View all (19) MYBBP1A antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MYBBP1A Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          MYBBP1A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MYBBP1A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MYBBP1A Antibody (H00010514-B01P) in WB

          MYBBP1A MaxPab polyclonal antibody. Western Blot analysis of MYBBP1A expression in 293. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MYBBP1A Antibody in Western Blot (WB)

          Product Details

          H00010514-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MYBBP1A (AAH50546.1, 1 a.a. approximately 1328 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Cell lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MESRDPAQPM SPGEATQSGA RPADRYGLLK HSREFLDFFW DIAKPEQETR LAATEKLLEY LRGRPKGSEM KYALKRLITG LGVGRETARP CYSLALAQLL QSFEDLPLCS ILQQIQEKYD LHQVKKAMLR PALFANLFGV LALFQSGRLV KDQEALMKSV KLLQALAQYQ NHLQEQPRKA LVDILSEVSK ATLQEILPEV LKADLNIILS SPEQLELFLL AQQKVPSKLK KLVGSVNLFS DENVPRLVNV LKMAASSVKK DRKLPAIALD LLRLALKEDK FPRFWKEVVE QGLLKMQFWP ASYLCFRLLG AALPLLTKEQ LHLVMQGDVI RHYGEHVCTA KLPKQFKFAP EMDDYVGTFL EGCQDDPERQ LAVLVAFSSV TNQGLPVTPT FWRVVRFLSP PALQGYVAWL RAMFLQPDLD SLVDFSTNNQ KKAQDSSLHM PERAVFRLRK WIIFRLVSIV DSLHLEMEEA LTEQVARFCL FHSFFVTKKP TSQIPETKHP FSFPLENQAR EAVSSAFFSL LQTLSTQFKQ APGQTQGGQP WTYHLVQFAD LLLNHSHNVT TVTPFTAQQR QAWDRMLQTL KELEAHSAEA RAAAFQHLLL LVGIHLLKSP AESCDLLGDI QTCIRKSLGE KPRRSRTKTI DPQEPPWVEV LVEILLALLA QPSHLMRQVA RSVFGHICSH LTPRALQLIL DVLNPETSED ENDRVVVTDD SDERRLKGAE DKSEEGEDNR SSESEEESEG EESEEEERDG DVDQGFREQL MTVLQAGKAL GGEDSENEEE LGDEAMMALD QSLASLFAEQ KLRIQARRDE KNKLQKEKAL RRDFQIRVLD LVEVLVTKQP ENALVLELLE PLLSIIRRSL RSSSSKQEQD LLHKTARIFT HHLCRARRYC HDLGERAGAL HAQVERLVQQ AGRQPDSPTA LYHFNASLYL LRVLKGNTAE GCVHETQEKQ KAGTDPSHMP TGPQAASCLD LNLVTRVYST ALSSFLTKRN SPLTVPMFLS LFSRHPVLCQ SLLPILVQHI TGPVRPRRQA CLLLQKTLSM REVRSCFEDP EWKQLMGQVL AKVTENLRVL GEAQTKAQHQ QALSSLELLN VLFRTCKHEK LTLDLTVLLG VLQGQQQSLQ QGAHSTGSSR LHDLYWQAMK TLGVQRPKLE KKDAKEIPSA TQSPISKKRK KKGFLPETKK RKKRKSEDGT PAEDGTPAAT GGSQPPSMGR KKRNRTKAKV PAQANGTPTT KSPAPGAPTR SPSTPAKSPK LQKKNQKPSQ VNGAPGSPTE PAGQKQHQKA LPKKGVLGKS PLSALARKKA RLSLVIRSPS LLQSGAKKKA QVRKAGKP

          Target Information

          The Myb family of transcription factors, which includes the structurally related A-, B-, and c-Myb genes, regulate differentiation, cellular growth and transactivating gene expression. c-Myb plays an essential role in controlling the proliferation and differentiation of hematopoietic cells, cellular levels of c-Myb decreasing as cells reach terminal differentiation. Myb-binding protein 1A (MYBBP1A) is a novel nuclear protein localized predominantly, but not exclusively, in nucleoli. Although initially isolated as a c-Myb-interacting protein, MYBBP1A is expressed ubiquitously, and associates with a number of different transcription factors. MYBBP1A associates with the aromatic hydrocarbon receptor (AhR), enhancing transactivation and increasing AhR-dependent gene expression. MYBBP1A is a powerful negative regulator of PPAR gamma coactivator 1 alpha (PGC-1 alpha) by p38 MAPK and also acts as a nucleocytoplasmic shuttling protein that utilizes CRM1-dependent and independent nuclear export pathways.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: MYB binding protein (P160) 1a; Myb-binding protein 1A; p53-activated protein-2

          View more View less

          Gene Aliases: MYBBP1A; P160; PAP2

          View more View less

          UniProt ID: (Human) Q9BQG0

          View more View less

          Entrez Gene ID: (Human) 10514

          View more View less

          Function(s)
          core promoter binding transcription corepressor activity DNA-directed DNA polymerase activity protein binding transcription factor binding sequence-specific DNA binding poly(A) RNA binding
          Process(es)
          osteoblast differentiation transcription, DNA-templated regulation of transcription, DNA-templated nucleocytoplasmic transport respiratory electron transport chain circadian regulation of gene expression cellular response to glucose starvation positive regulation of gene expression, epigenetic negative regulation of transcription, DNA-templated positive regulation of cell cycle arrest DNA biosynthetic process intrinsic apoptotic signaling pathway by p53 class mediator positive regulation of anoikis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0