Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MESRDPAQPM SPGEATQSGA RPADRYGLLK HSREFLDFFW DIAKPEQETR LAATEKLLEY LRGRPKGSEM KYALKRLITG LGVGRETARP CYSLALAQLL QSFEDLPLCS ILQQIQEKYD LHQVKKAMLR PALFANLFGV LALFQSGRLV KDQEALMKSV KLLQALAQYQ NHLQEQPRKA LVDILSEVSK ATLQEILPEV LKADLNIILS SPEQLELFLL AQQKVPSKLK KLVGSVNLFS DENVPRLVNV LKMAASSVKK DRKLPAIALD LLRLALKEDK FPRFWKEVVE QGLLKMQFWP ASYLCFRLLG AALPLLTKEQ LHLVMQGDVI RHYGEHVCTA KLPKQFKFAP EMDDYVGTFL EGCQDDPERQ LAVLVAFSSV TNQGLPVTPT FWRVVRFLSP PALQGYVAWL RAMFLQPDLD SLVDFSTNNQ KKAQDSSLHM PERAVFRLRK WIIFRLVSIV DSLHLEMEEA LTEQVARFCL FHSFFVTKKP TSQIPETKHP FSFPLENQAR EAVSSAFFSL LQTLSTQFKQ APGQTQGGQP WTYHLVQFAD LLLNHSHNVT TVTPFTAQQR QAWDRMLQTL KELEAHSAEA RAAAFQHLLL LVGIHLLKSP AESCDLLGDI QTCIRKSLGE KPRRSRTKTI DPQEPPWVEV LVEILLALLA QPSHLMRQVA RSVFGHICSH LTPRALQLIL DVLNPETSED ENDRVVVTDD SDERRLKGAE DKSEEGEDNR SSESEEESEG EESEEEERDG DVDQGFREQL MTVLQAGKAL GGEDSENEEE LGDEAMMALD QSLASLFAEQ KLRIQARRDE KNKLQKEKAL RRDFQIRVLD LVEVLVTKQP ENALVLELLE PLLSIIRRSL RSSSSKQEQD LLHKTARIFT HHLCRARRYC HDLGERAGAL HAQVERLVQQ AGRQPDSPTA LYHFNASLYL LRVLKGNTAE GCVHETQEKQ KAGTDPSHMP TGPQAASCLD LNLVTRVYST ALSSFLTKRN SPLTVPMFLS LFSRHPVLCQ SLLPILVQHI TGPVRPRRQA CLLLQKTLSM REVRSCFEDP EWKQLMGQVL AKVTENLRVL GEAQTKAQHQ QALSSLELLN VLFRTCKHEK LTLDLTVLLG VLQGQQQSLQ QGAHSTGSSR LHDLYWQAMK TLGVQRPKLE KKDAKEIPSA TQSPISKKRK KKGFLPETKK RKKRKSEDGT PAEDGTPAAT GGSQPPSMGR KKRNRTKAKV PAQANGTPTT KSPAPGAPTR SPSTPAKSPK LQKKNQKPSQ VNGAPGSPTE PAGQKQHQKA LPKKGVLGKS PLSALARKKA RLSLVIRSPS LLQSGAKKKA QVRKAGKP
The Myb family of transcription factors, which includes the structurally related A-, B-, and c-Myb genes, regulate differentiation, cellular growth and transactivating gene expression. c-Myb plays an essential role in controlling the proliferation and differentiation of hematopoietic cells, cellular levels of c-Myb decreasing as cells reach terminal differentiation. Myb-binding protein 1A (MYBBP1A) is a novel nuclear protein localized predominantly, but not exclusively, in nucleoli. Although initially isolated as a c-Myb-interacting protein, MYBBP1A is expressed ubiquitously, and associates with a number of different transcription factors. MYBBP1A associates with the aromatic hydrocarbon receptor (AhR), enhancing transactivation and increasing AhR-dependent gene expression. MYBBP1A is a powerful negative regulator of PPAR gamma coactivator 1 alpha (PGC-1 alpha) by p38 MAPK and also acts as a nucleocytoplasmic shuttling protein that utilizes CRM1-dependent and independent nuclear export pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MYB binding protein 1a; MYB binding protein (P160) 1a; Myb-binding protein 1A; p53-activated protein-2; PAR interacting protein; PAR-interacting protein; PIP
Gene Aliases: DBP; MYBBP1A; P160; PAP2
UniProt ID: (Human) Q9BQG0, (Rat) O35821
Entrez Gene ID: (Human) 10514, (Rat) 60571
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support