Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The Myb family of transcription factors, which includes the structurally related A-, B-, and c-Myb genes, regulate differentiation, cellular growth and transactivating gene expression. c-Myb plays an essential role in controlling the proliferation and differentiation of hematopoietic cells, cellular levels of c-Myb decreasing as cells reach terminal differentiation. Myb-binding protein 1A (MYBBP1A) is a novel nuclear protein localized predominantly, but not exclusively, in nucleoli. Although initially isolated as a c-Myb-interacting protein, MYBBP1A is expressed ubiquitously, and associates with a number of different transcription factors. MYBBP1A associates with the aromatic hydrocarbon receptor (AhR), enhancing transactivation and increasing AhR-dependent gene expression. MYBBP1A is a powerful negative regulator of PPAR gamma coactivator 1 alpha (PGC-1 alpha) by p38 MAPK and also acts as a nucleocytoplasmic shuttling protein that utilizes CRM1-dependent and independent nuclear export pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MYB binding protein 1a; MYB binding protein (P160) 1a; Myb-binding protein 1A; Myb-binding protein of 160 kDa; nuclear protein P160; p53-activated protein-2; PAR interacting protein; PAR-interacting protein; PIP
Gene Aliases: AL024407; AU019902; DBP; MYBBP1A; P160; p160MBP; p67MBP; PAP2
UniProt ID: (Human) Q9BQG0, (Rat) O35821, (Mouse) Q7TPV4
Entrez Gene ID: (Human) 10514, (Rat) 60571, (Mouse) 18432
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support