Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MGNSKGDRG NISTSSKPAS TSGKSELSSK HSRSLKPDGR MSRTTADQKK PRGTESLSAS ESLILKSDAA KLRSDSHSRS LSPNHNTLQT LKSDGRMPSS SRAESPGPGS RLSSPKPKTL PANRSSPSGA SSPRSSSPHD KNLPQKSTAP VKTKLDPPRE RSKSDSYTLD PDTLRKKKMP LTEPLRGRST SPKPKSVPKD STDSPGSENR A (2672-2880 aa encoded by NM _015057)
This gene encodes an E3 ubiquitin-protein ligase and member of the PHR (Phr1/MYCBP2, highwire and RPM-1) family of proteins. The encoded protein plays a role in axon guidance and synapse formation in the developing nervous system. In mammalian cells, this protein regulates the cAMP and mTOR signaling pathways, and may additionally regulate autophagy. Reduced expression of this gene has been observed in acute lymphoblastic leukemia patients and a mutation in this gene has been identified in patients with a rare inherited vision defect.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase MYCBP2; Highwire; Myc-binding protein 2; pam/highwire/rpm-1 protein; probable E3 ubiquitin-protein ligase MYCBP2; Protein associated with Myc
Gene Aliases: KIAA0916; Myc-bp2; MYCBP2; PAM; Phr
UniProt ID: (Human) O75592
Entrez Gene ID: (Human) 23077
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support