Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDGPGFGGM NRIGGGIGFG GLEAMNSMGG FGGVGRMGEL YRGAMTSSME RDFGRGDIGI NRGFGDSFGR LGGGMGSMNS VTGGMGMGLD RMSSSFDRMG PGIGAILERS IDMDRGFLSG PMGSGMRERI GSKGNQIFVR NLPFDLTWQK LKEKFSQCGH VMFAEIKMEN GKSKGCGTVR FDSPESAEKA CRIMNGIKIS GREIDVRLDR NA (1-212 aa encoded by BC014533)
MEF-2 is expressed early in the differentiation program and is suppressed by specific polypeptide growth factors. The ability of MEF-2 to recognize conserved activating elements associated with multiple-specific genes suggests that this factor may participate in the coordinate regulation of genes during myogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MEF-2; MST156; myEF-2; myelin basic protein expression factor 2, repressor; Myelin expression factor 2; myelin gene expression factor 2
Gene Aliases: 9430071B01; HsT18564; KIAA1341; MEF-2; Mef2; mKIAA1341; MST156; MSTP156; myEF-2; MYEF2
UniProt ID: (Human) Q9P2K5, (Mouse) Q8C854
Entrez Gene ID: (Human) 50804, (Rat) 679712, (Mouse) 17876
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support