Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • MYH6/MYH7 Antibodies

          Bioss

          MYH Polyclonal Antibody

          View all (2) MYH6/MYH7 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MYH Polyclonal Antibody

          Product Details

          BS-15444R

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:500
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -

          Flow Cytometry (Flow)

          Assay-dependent
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human MYH 6.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          Cardiac muscle myosin is a hexamer consisting of two heavy chain subunits, two light chain subunits, and two regulatory subunits. This gene encodes the alpha heavy chain subunit of cardiac myosin. The gene is located 4kb downstream of the gene encoding the beta heavy chain subunit of cardiac myosin. Mutations in this gene cause familial hypertrophic cardiomyopathy and atrial septal defect 3.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: alpha cardiac MHC; alpha myosin; beta myosin heavy chain; cardiac muscle myosin heavy chain 7 beta; cardiac myosin heavy chain alpha isoform; cardiac myosin heavy chain beta isoform; cardiomyopathy, hypertrophic 1; MYH-beta/slow; MyHC-alpha; MyHC-beta; MyHC-slow; myopathy, distal 1; Myosin heavy chain 6; Myosin heavy chain 7; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain polypeptide 7 cardiac muscle fetal; Myosin heavy chain slow isoform; Myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); Myosin heavy chain, cardiac muscle beta isoform; myosin heavy chain, cardiac muscle, adult; myosin heavy chain, cardiac muscle, fetal; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin heavy chain, polypeptide 7; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin, heavy polypeptide 7, cardiac muscle, beta; Myosin-6; Myosin-7; rhabdomyosarcoma antigen MU-RMS-40.7A

          View more View less

          Gene Aliases: A830009F23Rik; AA517445; alpha-MHC; ASD3; B-MHC; beta-MHC; CMD1EE; CMD1S; CMH1; CMH14; MPD1; MYH6; MYH7; MYHC; Myhc-a; Myhc-b; MyHC-I; MYHCA; MYHCB; SPMD; SPMM; SSS3

          View more View less

          UniProt ID: (Human) P13533, (Human) P12883, (Mouse) Q02566, (Mouse) Q91Z83, (Rat) P02564

          View more View less

          Entrez Gene ID: (Human) 4624, (Human) 4625, (Mouse) 17888, (Rat) 29556, (Mouse) 140781, (Rat) 29557

          View more View less

          Function(s)
          microfilament motor activity actin binding calmodulin binding ATP binding ATPase activity myosin phosphatase activity protein kinase binding actin-dependent ATPase activity protein binding protein homodimerization activity protein heterodimerization activity nucleotide binding motor activity calcium-dependent ATPase activity identical protein binding
          Process(es)
          in utero embryonic development regulation of the force of heart contraction regulation of heart rate muscle contraction striated muscle contraction adult heart development visceral muscle development regulation of heart contraction metabolic process regulation of blood pressure actin filament-based movement muscle filament sliding myofibril assembly BMP signaling pathway regulation of ATPase activity sarcomere organization ATP metabolic process cardiac muscle fiber development atrial cardiac muscle tissue morphogenesis ventricular cardiac muscle tissue morphogenesis cardiac muscle contraction canonical Wnt signaling pathway regulation of heart growth response to heat skeletal muscle contraction regulation of the force of skeletal muscle contraction transition between fast and slow fiber regulation of slow-twitch skeletal muscle fiber contraction response to reactive oxygen species protein dephosphorylation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67c77f6dc7-n6nf2:80/100.66.135.186:80.
          git-commit: 545e792480003146e539d624bd2a21b51d198228
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0