Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Cardiac muscle myosin is a hexamer consisting of two heavy chain subunits, two light chain subunits, and two regulatory subunits. This gene encodes the alpha heavy chain subunit of cardiac myosin. The gene is located 4kb downstream of the gene encoding the beta heavy chain subunit of cardiac myosin. Mutations in this gene cause familial hypertrophic cardiomyopathy and atrial septal defect 3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiac muscle myosin heavy chain 7 beta; MyHC-alpha; MyHC-beta; MyHC-slow; myopathy, distal 1; Myosin heavy chain 6; Myosin heavy chain 7; Myosin heavy chain slow isoform; Myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle beta isoform; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin, heavy polypeptide 7, cardiac muscle, beta; Myosin-6; Myosin-7; rhabdomyosarcoma antigen MU-RMS-40.7A
Gene Aliases: alpha-MHC; ASD3; CMD1EE; CMD1S; CMH1; CMH14; MPD1; MYH6; MYH7; MYHC; MYHCA; MYHCB; SPMD; SPMM; SSS3
UniProt ID: (Human) P13533, (Human) P12883
Entrez Gene ID: (Human) 4624, (Human) 4625
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support