Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MQARRGKGA WEKTRLALEA EVSELRAELS SLQTARQEGE QRRRRLELQL QEVQGRAGDG ERARAEAAEK LQRAQAELEN VSGALNEAES KTIRLSKELS STEAQLHDAQ ELLQEETRAK LALGSRVRAM EAEAAGLREQ LEEEAAARER AGRELQTAQA QLSEWRRRQE EEAGALEAGE EARRRAAREA EALTQRLAEK TE (1231-1430 aa encoded by NM_001077186)
Cellular myosin that appears to play a role in cytokinesis, cell shape, and specialized functions such as secretion and capping.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MYH14 variant protein; Myosin heavy chain 14; Myosin heavy chain, non-muscle IIc; myosin, heavy chain 14; myosin, heavy polypeptide 14; Myosin-14; NMHC II-C; Non-muscle myosin heavy chain IIc; nonmuscle myosin heavy chain; nonmuscle myosin heavy chain II-C
Gene Aliases: 2400004E04Rik; DFNA4; DFNA4A; FP17425; II-C; KIAA2034; MHC16; MYH14; MYH17; myosin; NHMCII; NMHC II-C; NMHC-II-C; PNMHH
UniProt ID: (Human) Q7Z406, (Mouse) Q6URW6
Entrez Gene ID: (Human) 79784, (Mouse) 71960
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support