Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MATENGAVE LGIQNPSTDK APKGPTGERP LAAGKDPGPP DPKKAPDPPT LKKDAKAPAS EKGDGTLAQP STSSQGPKGE GDRGGGPAEG SAGPPAALPQ QTATPETSVK KPKAEQGASG SQDPGKPRVG KKAAEGQAAA RRGSPAFLHS PSCPAIISSS EKLLAKKPPS EASELTFEGV PMTHSPTDPR PAKAEEGKNI LAESQKEVGE KTPGQAGQAK MQGDTSRGIE FQAVPSEKSE VGQALCLTAR EEDCFQILDD CPPPPAPFPH RMVELRTGNV SSEFSMNSKE ALGGGKFGAV C (1-300 aa encoded by BC069627)
MYLK2 (skMLCK) is a calcium/calmodulin-dependent serine/threonine kinase expressed exclusively in adult skeletal muscle. MYLK2 encodes a myosin light chain kinase, a calcium/calmodulin dependent enzyme, that is exclusively expressed in adult skeletal muscle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MLCK2; myosin light chain kinase 2, skeletal muscle; Myosin light chain kinase 2, skeletal/cardiac muscle
Gene Aliases: 9830004H17Rik; BB138278; KMLC; MLCK; MLCK2; MYLK2; skMLCK
UniProt ID: (Human) Q9H1R3, (Mouse) Q8VCR8
Entrez Gene ID: (Human) 85366, (Mouse) 228785
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support