Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MATENGAVEL GIQNPSTDKA PKGPTGERPL AAGKDPGPPD PKKAPDPPTL KKDAKAPASE KGDGTLAQPS TSSQGPKGEG DRGGGPAEGS AGPPAALPQQ TATPETSVKK PKAEQGASGS QDPGKPRVGK KAAEGQAAAR RGSPAFLHSP SCPAIISSSE KLLAKKPPSE ASELTFEGVP MTHSPTDPRP AKAEEGKNIL AESQKEVGEK TPGQAGQAKM QGDTSRGIEF QAVPSEKSEV GQALCLTARE EDCFQILDDC PPPPAPFPHR MVELRTGNVS SEFSMNSKEA LGGGKFGAVC TCMEKATGLK LAAKVIKKQT PKDKEMVLLE IEVMNQLNHR NLIQLYAAIE TPHEIVLFME YIEGGELFER IVDEDYHLTE VDTMVFVRQI CDGILFMHKM RVLHLDLKPE NILCVNTTGH LVKIIDFGLA RRYNPNEKLK VNFGTPEFLS PEVVNYDQIS DKTDMWSMGV ITYMLLSGLS PFLGDDDTET LNNVLSGNWY FDEETFEAVS DEAKDFVSNL IVKDQRARMN AAQCLAHPWL NNLAEKAKRC NRRLKSQILL KKYLMKRRWK KNFIAVSAAN RFKKISSSGA LMALGV
MYLK2 (skMLCK) is a calcium/calmodulin-dependent serine/threonine kinase expressed exclusively in adult skeletal muscle. MYLK2 encodes a myosin light chain kinase, a calcium/calmodulin dependent enzyme, that is exclusively expressed in adult skeletal muscle.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MLCK2; myosin light chain kinase 2, skeletal muscle; Myosin light chain kinase 2, skeletal/cardiac muscle
Gene Aliases: KMLC; MLCK; MLCK2; MYLK2; skMLCK
UniProt ID: (Human) Q9H1R3
Entrez Gene ID: (Human) 85366
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support