Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • MYST1 Antibodies

          Abnova

          MYST1 Polyclonal Antibody, MaxPab™

          View all (16) MYST1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MYST1 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          MYST1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MYST1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MYST1 Antibody (H00084148-D01P) in WB

          Western Blot analysis of MYST1 expression in transfected 293T cell line (H00084148-T01) by MYST1 MaxPab polyclonal antibody.

          Lane 1: MYST1 transfected lysate(53.10 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MYST1 Antibody in Western Blot (WB)

          Product Details

          H00084148-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MYST1 (AAH37773.1, 1 a.a. approximately 467 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MAAQGAAAAV AAGTSGVAGE GEPGPGENAA AEGTAPSPGR VSPPTPARGE PEVTVEIGET YLCRRPDSTW HSAEVIQSRV NDQEGREEFY VHYVGFNRRL DEWVDKNRLA LTKTVKDAVQ KNSEKYLSEL AEQPERKITR NQKRKHDEIN HVQKTYAEMD PTTAALEKEH EAITKVKYVD KIHIGNYEID AWYFSPFPED YGKQPKLWLC EYCLKYMKYE KSYRFHLGQC QWRQPPGKEI YRKSNISVYE VDGKDHKIYC QNLCLLAKLF LDHKTLYFDV EPFVFYILTE VDRQGAHIVG YFSKEKESPD GNNVACILTL PPYQRRGYGK FLIAFSYELS KLESTVGSPE KPLSDLGKLS YRSYWSWVLL EILRDFRGTL SIKDLSQMTS ITQNDIISTL QSLNMVKYWK GQHVICVTPK LVEEHLKSAQ YKKPPITGGW GAAVCRGRWG SVSMWTGRSQ GLLIAVT

          Target Information

          The MYST family of histone acetyltransferases, which includes MYST1, is named for the founding members MOZ (MYST3), yeast YBF2 and SAS2, and TIP60 (HTATIP). All members of this family contain a MYST region of about 240 amino acids with a canonical acetyl-CoA-binding site and a C2HC-type zinc finger motif. Most MYST proteins also have a chromodomain involved in protein-protein interactions and targeting transcriptional regulators to chromatin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: h FLJ14040; Histone acetyltransferase KAT8; histone acetyltransferase MYST1; hMOF; K(lysine) acetyltransferase 8; Lysine acetyltransferase 8; Males-absent on the first protein homolog; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 1; MYST histone acetyltransferase 1; MYST-1; ortholog of Drosophila males absent on the first (MOF); probable histone acetyltransferase MYST1; Protein acetyltransferase KAT8; Protein propionyltransferase KAT8

          View more View less

          Gene Aliases: hMOF; KAT8; MOF; MYST1; PP7073; ZC2HC8

          View more View less

          UniProt ID: (Human) Q9H7Z6

          View more View less

          Entrez Gene ID: (Human) 84148

          View more View less

          Function(s)
          histone acetyltransferase activity protein binding transcription factor binding acetyltransferase activity enzyme binding methylated histone binding histone acetyltransferase activity (H4-K5 specific) histone acetyltransferase activity (H4-K8 specific) metal ion binding histone acetyltransferase activity (H4-K16 specific)
          Process(es)
          transcription, DNA-templated regulation of autophagy histone acetylation myeloid cell differentiation histone H4-K5 acetylation histone H4-K8 acetylation histone H4-K16 acetylation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0