Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody recognizes porcine cells of the monocyte/macrophage lineage. Expression of the antigen is increased with maturation, with higher expression on peritoneal and alveolar macrophages. Some expression has also been observed on peripheral blood lymphocytes.
Macrophage antibody recognizes porcine cells of the monocyte/macrophage lineage. Expression of the antigen is increased with maturation, with higher expression on peritoneal and alveolar macrophages.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calprotectin; protein S100-A8
Entrez Gene ID: (Pig) 100127488
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support