Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Macrophages (Mature) Antibodies

          Invitrogen

          Macrophages (Mature) Monoclonal Antibody (25F9), Biotin

          View all (3) Macrophages (Mature) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Macrophages (Mature) Monoclonal Antibody (25F9), Biotin

          Product Details

          MA5-49062

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:50
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:50
          -

          Flow Cytometry (Flow)

          1:50
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          25F9

          Immunogen

          The monoclonal antibody 25F9 recognizes a protein of 86 kD on the cell surface and within the cytoplasm of mature macrophages.

          Conjugate

          Biotin Biotin Biotin
          • Unconjugated
          • FITC
          • Request custom conjugation

          Form

          Liquid

          Concentration

          100 µg/mL

          Purification

          Protein G

          Storage buffer

          PBS with 0.1% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3090506

          Product Specific Information

          The antibody is associated with fully differentiated tissue macrophages both in normal and in diseased tissues, independently of the presence or absence of inflammation. The antigen is absent on freshly isolated monocytes and other blood cells. After 6 to 7 days culture human monocytes become positive. Some melanoma and carcinoma cell lines are also positive. Furthermore the monoclonal antibody 25F9 cross reacts with a subpopulation of macrophages of rhesus monkey, pig alveolar macrophages and Kupffer cells.

          Target Information

          Macrophages are a type of white blood cell that plays a crucial role in the immune system. They are responsible for detecting, engulfing, and destroying foreign substances, such as bacteria and viruses, as well as dead cells and debris. Mature macrophages are found in various tissues throughout the body and are highly specialized to perform specific functions depending on their location. For example, alveolar macrophages are found in the lungs and are responsible for removing foreign particles that are inhaled, while Kupffer cells are found in the liver and are responsible for removing old or damaged red blood cells. Understanding the function and behavior of macrophages is crucial for developing new treatments for diseases such as cancer, autoimmune disorders, and infections.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: casein kinase 1, alpha 1; casein kinase 1, alpha 1-like; casein kinase I alpha S-like; Casein kinase I isoform alpha; Casein kinase I isoform alpha-like; CASP-1; CASP1 nirs variant 1; caspase 1, apoptosis-related cysteine peptidase (interleukin 1, beta, convertase); Caspase-1; CK1; CKI-alpha; CKI-alpha-like; clock regulator kinase; down-regulated in lung cancer; epididymis secretory sperm binding protein Li 77p; ICE; IL-1 beta-converting enzyme; IL-1BC; IL1B-convertase; interleukin 1, beta, convertase; interleukin 1-B converting enzyme; Interleukin-1 beta convertase; Interleukin-1 beta-converting enzyme; Macrophages; Mature Macrophages; p45

          View more View less

          Gene Aliases: CASP1; CK1; CK1a; CKIa; CSNK1A1; CSNK1A1L; HEL-S-77p; HLCDGP1; ICE; IL1BC; IL1BCE; P45; PRO2975

          View more View less

          UniProt ID: (Human) P29466, (Human) P48729, (Human) Q8N752

          View more View less

          Entrez Gene ID: (Human) 834, (Human) 1452, (Human) 122011

          View more View less

          Function(s)
          endopeptidase activity cysteine-type endopeptidase activity protein binding cysteine-type endopeptidase activator activity involved in apoptotic process cysteine-type endopeptidase activity involved in apoptotic process protein kinase activity protein serine/threonine kinase activity ATP binding kinase activity
          Process(es)
          response to hypoxia proteolysis apoptotic process activation of cysteine-type endopeptidase activity involved in apoptotic process signal transduction regulation of autophagy protein processing response to lipopolysaccharide interleukin-1 beta production response to ATP regulation of apoptotic process positive regulation of I-kappaB kinase/NF-kappaB signaling positive regulation of interleukin-1 alpha secretion positive regulation of interleukin-1 beta secretion regulation of inflammatory response mitochondrial depolarization membrane hyperpolarization pyroptosis cellular response to mechanical stimulus cellular response to organic substance programmed necrotic cell death toxin transport cell morphogenesis phagocytosis regulation of cell shape Wnt signaling pathway peptidyl-serine phosphorylation protein phosphorylation Golgi organization mitotic nuclear division cell surface receptor signaling pathway positive regulation of proteasomal ubiquitin-dependent protein catabolic process proteasome-mediated ubiquitin-dependent protein catabolic process intermediate filament cytoskeleton organization cell division negative regulation of canonical Wnt signaling pathway regulation of GTP binding beta-catenin destruction complex assembly beta-catenin destruction complex disassembly
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0