Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Mammaglobin is a gene that is expressed almost exclusively in the normal breast epithelium and human breast cancer. It is a member of the secretoglobin gene family and forms a heterodimer with lipophilin B. It has been suggested that mammaglobin may be a useful marker for breast cancer clinical research. Studies investigating the detection of mRNA by RT PCR from circulating carcinoma cells in the peripheral blood of breast cancer patients have shown that mammaglobin is a highly specific marker and correlates with several prognostic factors, such as lymph node involvement.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C3.1; HGNC:7050; mammaglobin A; MGC71974; prostatein; prostatein C3 subunit; Prostatein peptide C3; prostatic steroid binding protein, allele C3(1); Prostatic steroid-binding protein C3; Secretoglobin family 2A member 2
Gene Aliases: Psbpc3; Scgb2a2
UniProt ID: (Rat) P02780
Entrez Gene ID: (Rat) 361725
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support