Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody detects the Myosin heavy chain of drosophila.
Immunogen sequence: RNQLFQQKK KADQEISGLK KDIEDLELNV QKAEQDKATK DHQIRNLNDE IAHQDELINK LNKEKKMQGE TNQKTGEELQ AAEDKINHLN KVKAKLEQTL DELEDSLERE KKVRGDVEKS KRKVEGDLKL TQEAVADLER NKKELEQTIQ RKDKELSSIT AKLEDEQVVV LKHQRQIKEL QARIE (932-1115 aa encoded by NM_078863 )
The function of Mhc is muscle contraction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: beta-myosin heavy chain; CG17927-PA; CG17927-PB; CG17927-PC; CG17927-PD; CG17927-PE; CG17927-PF; CG17927-PG; CG17927-PH; CG17927-PI; CG17927-PK; CG17927-PL; CG17927-PM; CG17927-PN; CG17927-PO; CG17927-PP; CG17927-PQ; CG17927-PR; CG17927-PS; CG17927-PT; CG17927-PU; CG17927-PV; chain; heavy chain; indirect flight muscle (2) RU1; indirect flight muscle (2) RU2; Mhc-PA; Mhc-PB; Mhc-PC; Mhc-PD; Mhc-PE; Mhc-PF; Mhc-PG; Mhc-PH; Mhc-PI; Mhc-PK; Mhc-PL; Mhc-PM; Mhc-PN; Mhc-PO; Mhc-PP; Mhc-PQ; Mhc-PR; Mhc-PS; Mhc-PT; Mhc-PU; Mhc-PV; muscle myosin; muscle myosin heavy chain; muscle myosin II; muscle myosin II heavy chain; muscle myosin-II; muscle specific myosin II; myo-II heavy chain; myosin; myosin heavy; myosin heavy chain; Myosin heavy chain, muscle; myosin heavy Chain-PF; myosin heavy-chain; myosin heavy-chain protein; myosin rod protein; Not-upheld; samba; sarcomeric myosin heavy-chain; shrunken thorax; Shrunken-thorax; stuckup
Gene Aliases: Bsh; CG17927; chr2L:16765388..16765564; chr2L:16781880..16782040; Dm II; Dmel\CG17927; Dmel_CG17927; DmMHC; DROMHC; DroMII; FBgn0002741; Ifm(2)2; Ifm(2)RU1; ifm(2)RU2; l(2)36Ae; l(2)k10423; l(2)M66; MHC; Mhc36B; mMHC; MRP; MyHC; Myo; Nup; Sht; sMHC; Stp
UniProt ID: (Fruit fly) P05661
Entrez Gene ID: (Fruit fly) 35007
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support