Invitrogen
This antibody is specific to M. genitalium and does not cross-react with Mycoplasma hominis, Mycoplasma pneumoniae, Ureaplasma urealyticum, Herpes virus type 2, Chlamydia trachomatis, Neisseria gonorrhoeae, or human cells.
Purity is >90%.
Recommended pairs for ELISA: MA5-18228 (capture) and MA5-18227 (conjugated, detection).