Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • MYL9/MYL12 Antibodies

          Rockland

          Myosin Polyclonal Antibody

          View all (8) MYL9/MYL12 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Myosin Polyclonal Antibody

          • Antibody Testing Data (1)
          Myosin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Myosin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Myosin Antibody (600-401-938) in WB

          Western blot using Rocklands anti-RLC of Smooth and Non-muscle Myosin antibody to detect vascular myosin (rat aorta, lane 1) but not cardiac myosin (mouse heart, lane2). Each lane was loaded with 35 µg of lysate. Arrowheads indicate the detection of both mono-phosphorylated (upper) and unphosphorylated (lower) forms of the protein. After blocking with 5% Normal goat serum and 0.5% BLOTTO in PBS, the membrane was probed with the primary antibody diluted in blocking buffer to 1:600 for 2 h at room temperature. The membrane was washed and r... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Myosin Antibody in Western Blot (WB)

          Product Details

          600-401-938

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:250-1:1,000
          -

          Immunohistochemistry (IHC)

          Assay-dependent
          -

          ELISA (ELISA)

          1:10,000-1:70,000
          -

          Immunoprecipitation (IP)

          1:500-1:2,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          This affinity purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids aa 10-35 of human myosin light chain protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.98 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          0.02M potassium phosphate, pH 7.2, with 0.15M NaCl

          Contains

          0.01% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Store vial at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4° C as an undiluted liquid. Dilute only prior to immediate use.

          This affinity purified antibody is directed against the regulatory light chain of smooth and non-muscle myosin. The antibody detects both unphosphorylated and monophosphorylated forms of the protein. Cross reactivity is expected with myosin light chain from human, mouse and rat sources. Reactivity with the protein from other species has not been determined; however, the sequence of the immunogen is nearly identical in mammalian and avian species. BLAST search analysis was used to determine that the smooth and non-muscle forms of myosin regulatory light chain have identical sequences. Cross reactivity is expected.

          Target Information

          Myosin regulatory light polypeptide 9 is a highly conserved, ubiquitously expressed protein that in humans is encoded by the MYL9 gene. Myosin, a structural component of muscle, are hexameric proteins consisting of two heavy chains and four light chains, including a single pair of nonphosphorylatable light chain subunits and another pair of phosphorylatable light chain subunits.The protein encoded by this gene is a myosin light chain that may regulate muscle contraction by modulating the ATPase activity of myosin heads. Myosin light chain (Myl) 9 and Myl12 are new functional ligands for CD69. Blockade of CD69-Myl9/12 interactio ameliorates allergic airway inflammation in ovalbumin-induced and house dust mite-induced mouse models of asthma.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 20 kDa myosin light chain; brain specific myosin regulatory light chain; Epididymis secretory protein Li 24; HEL-S-24; LC20; MLC-2; MLC-2A; MLC-2B; MLC-2C; MLC20; myosin light chain, regulatory B; myosin light chain, regulatory B-like; myosin light chain, regulatory C; myosin regulatory light chain 1; Myosin regulatory light chain 12A; Myosin regulatory light chain 12B; myosin regulatory light chain 2; Myosin regulatory light chain 2, nonsarcomeric; Myosin regulatory light chain 2, smooth muscle isoform; Myosin regulatory light chain 2-A, smooth muscle isoform; Myosin regulatory light chain 2-B, smooth muscle isoform; Myosin regulatory light chain 20 kDa; myosin regulatory light chain 3; Myosin regulatory light chain 9; Myosin regulatory light chain MRLC1; Myosin regulatory light chain MRLC2; Myosin regulatory light chain MRLC3; Myosin regulatory light chain RLC-A; myosin regulatory light chain-B; Myosin regulatory light polypeptide 9; Myosin RLC; Myosin RLC-A; Myosin RLC-B; myosin, light chain 12A, regulatory, non-sarcomeric; myosin, light chain 12B, regulatory; myosin, light chain 9, regulatory; myosin, light polypeptide 9, regulatory; myosin, light polypeptide, regulatory, non-sarcomeric (20kD); NMDA receptor-interacting protein; SHUJUN-1

          View more View less

          Gene Aliases: 1500001M02Rik; 2900073G15Rik; AI327049; AI838292; C77744; C80200; HEL-S-24; LC20; MLC-2B; MLC-2C; MLC-B; MLC2; MLC20; MLCB; MRCL3; MRLC1; MRLC2; MRLC3; Mrlcb; MYL12A; MYL12B; MYL2B; MYL9; MYLC2B; Mylc2c; MYRL2; RGD1309537; RLC; Rlc-a; RLC-B; RLC-C

          View more View less

          UniProt ID: (Human) P19105, (Human) O14950, (Human) P24844, (Rat) P13832, (Rat) P18666, (Mouse) Q3THE2, (Rat) Q64122, (Mouse) Q9CQ19

          View more View less

          Entrez Gene ID: (Human) 10627, (Human) 103910, (Human) 10398, (Rat) 501203, (Mouse) 67268, (Rat) 50685, (Mouse) 67938, (Rat) 296313, (Mouse) 98932

          View more View less

          Function(s)
          calcium ion binding protein binding myosin heavy chain binding structural constituent of muscle glutamate receptor binding metal ion binding
          Process(es)
          platelet aggregation regulation of cell shape protein targeting to plasma membrane muscle contraction ephrin receptor signaling pathway regulation of muscle contraction
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0