Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Additional Information: This antibody is expected to recognize both reported isoforms (NP_006781.1; NP_001129412.1).
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
This gene encodes a cystoskeletal protein which plays a significant role in the stability of thin filaments during muscle contraction. This protein binds F-actin, crosslinks actin filaments, and prevents latrunculin A-induced filament disassembly. Mutations in this gene have been associated with limb-girdle muscular dystrophy and myofibrillar myopathies. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 57 kDa cytoskeletal protein; Myofibrillar titin-like Ig domains protein; Myotilin; Titin immunoglobulin domain protein; titin immunoglobulin domain protein (myotilin)
Gene Aliases: LGMD1; LGMD1A; MFM3; MYOT; TTID; TTOD
UniProt ID: (Human) Q9UBF9
Entrez Gene ID: (Human) 9499
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support