Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ADEKEDLPNS STSFLFDKIP KQEEKLLPEL SSNHIIPNIQ DTQVHLGVSD DLGLLAHMTG SVDLTCTSSI IEEKEFKIHT TSDGMSSI
This gene encodes a member of a large, diverse group of A-kinase anchor proteins that target the activity of protein kinase A to specific subcellular sites by binding to its type II regulatory subunits. Brain-specific expression and coat protein-like membrane recruitment of a highly similar protein in mouse suggest an involvement in neuronal post-Golgi membrane traffic. Mutations in this gene may be associated with a form of autism. This gene and its expression are frequently disrupted in patients with multiple myeloma. Alternative splicing results in multiple transcript variants encoding distinct isoforms. Additional transcript variants may exist, but their full-length nature has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ10197; Lysosomal-trafficking regulator 2; Neurobeachin; Protein BCL8B; RP11-270C18.1
Gene Aliases: BCL8B; KIAA1544; LYST2; NBEA
UniProt ID: (Human) Q8NFP9
Entrez Gene ID: (Human) 26960
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support