Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is specific to NCALD.
Immunogen sequence: MGKQNSKLR PEVMQDLLES TDFTEHEIQE WYKGFLRDCP SGHLSMEEFK KIYGNFFPYG DASKFAEHVF RTFDANGDGT IDFREFIIAL SVTSRGKLEQ KLKWAFSMYD LDGNGYISKA EMLEIVQAIY KMVSSVMKMP EDESTPEKRT EKIFRQMDTN RDGKLSMEEF IRGAKSDPSI VRLLQCDPSS AGQF (1-193 aa encoded by BC036098)
This gene encodes a member of the neuronal calcium sensor (NCS) family of calcium-binding proteins. The protein contains an N-terminal myristoylation signal and four EF-hand calcium binding loops. The protein is cytosolic at resting calcium levels; however, elevated intracellular calcium levels induce a conformational change that exposes the myristoyl group, resulting in protein association with membranes and partial co-localization with the perinuclear trans-golgi network. The protein is thought to be a regulator of G protein-coupled receptor signal transduction. Several alternatively spliced variants of this gene have been determined, all of which encode the same protein; additional variants may exist but their biological validity has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neurocalcin delta; Neurocalcin-delta
Gene Aliases: AI848120; D030020D09Rik; D15Ertd412e; NCALD
UniProt ID: (Human) P61601, (Rat) Q5PQN0, (Mouse) Q91X97
Entrez Gene ID: (Pig) 100156785, (Human) 83988, (Rat) 553106, (Mouse) 52589
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support