Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 3
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:4,000.
NCAM2 (neural cell adhesion molecule 2) is a cell adhesion molecule belonging to the immunoglobulin superfamily. It is differentially expressed in the developing and mature olfactory epithelium and is involved in the guidance of developing olfactory sensory neurons to the glomeruli of the olfactory bulb.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N-CAM-2; NCAM-2; Neural cell adhesion molecule 2; Neural cell adhesion molecule RB-8; olfactory axon cell adhesion molecule; R4B12; R4B12 antigen
Gene Aliases: Ncam-2; NCAM2; NCAM21; Ocam; RNCAM
UniProt ID: (Human) O15394, (Mouse) O35136
Entrez Gene ID: (Human) 4685, (Mouse) 17968
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support