Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • NCAPG Antibodies

          Proteintech

          NCAPG Monoclonal Antibody (2D4D3), CoraLite® Plus 488

          View all (12) NCAPG antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite NCAPG Monoclonal Antibody (2D4D3), CoraLite® Plus 488

          • Antibody Testing Data (2)
          NCAPG Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NCAPG Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NCAPG Antibody (CL488-68082) in ICC/IF

          Immunofluorescent analysis of (-20°C Methanol) fixed U2OS cells using CoraLite® Plus 488 NCAPG antibody (CL488-68082, Clone: 2D4D3 ) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NCAPG Antibody in Immunocytochemistry (ICC/IF)
          NCAPG Antibody in Flow Cytometry (Flow)

          Product Details

          CL488-68082

          Applications
          Tested Dilution
          Publications

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -

          Flow Cytometry (Flow)

          0.4 µg/1x10^6 cell suspension
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2D4D3

          Immunogen

          Recombinant protein
          View immunogen

          Conjugate

          CoraLite® Plus 488 CoraLite® Plus 488 CoraLite® Plus 488
          • Unconjugated
          • CoraLite 594
          • Request custom conjugation

          Excitation/Emission Max

          493/522 nm

          Form

          Liquid

          Purification

          Protein A

          Storage buffer

          PBS with 50% glycerol, 0.5% BSA

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, store in dark

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: TLHCEGTEIN SDDEQESKEV EETATAKNVL KLLSDFLDSE VSELRTGAAE GLAKLMFSGL LVSSRILSRL ILLWYNPVTE EDVQLRHCLG VFFPVFAYAS RTNQECFEEA FLPTLQTLAN APASSPLAEI DITNVAELLV DLTRPSGLNP QAKTSQDYQA LTVHDNLAMK ICNEILTSPC SPEIRVYTKA LSSLELSSHL AKDLLVLLNE ILEQVKDRTC LRALEKIKIQ LEKGNKEFGD QAEAAQDATL TTTTFQNEDE KNKEVYMTPL RGVKATQASK STQLKTNRGQ RKVTVSARTN RRCQTAEADS ESDHEVPEPE SEMKMRLPRR AKTAALEKSK LNLAQFLNED LS

          Target Information

          This gene encodes a subunit of the condensin complex, which is responsible for the condensation and stabilization of chromosomes during mitosis and meiosis. Phosphorylation of the encoded protein activates the condensin complex. There are pseudogenes for this gene on chromosomes 8 and 15. Alternative splicing results in multiple transcript variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: chromosome condensation protein G; Chromosome-associated protein G; Condensin complex subunit 3; Condensin subunit CAP-G; FLJ12450; hCAP-G; Melanoma antigen NY-MEL-3; MGC126525; Non-SMC condensin I complex subunit G; non-SMC condensin I complex, subunit G; XCAP-G homolog

          View more View less

          Gene Aliases: CAPG; CHCG; NCAPG; NY-MEL-3; NYMEL3; YCG1

          View more View less

          UniProt ID: (Human) Q9BPX3

          View more View less

          Entrez Gene ID: (Human) 64151

          View more View less

          Function(s)
          protein binding
          Process(es)
          mitotic chromosome condensation cell division
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-587cc6cfbd-ct8xs:80/100.66.129.252:80.
          git-commit: 43c1f9e84613abe662391d2f5aa289917b685966
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0