Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen Sequence: LRPHSALLEN MHIEQLARRL PARVQGYPWR LAYSTLEHGT SLKTLYRKSA SLDSPVLLVI KDMDNQIFGA YATHPFKFSD HYYGTGETFL YTFSPHFKVF KWSGENSYFI NGDISSLELG GGGGRFGLWL DADLYHGRSN SCSTFNNDIL SKKEDFIVQD LEVWAFD
NCOA7 enhances the transcriptional activities of several nuclear receptors. It is involved in the coactivation of different nuclear receptors, such as ESR1, THRB, PPARG and RARA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 140 kDa estrogen receptor-associated protein; Estrogen nuclear receptor coactivator 1; estrogen receptor associated protein 140 kDa; Nuclear receptor coactivator 7; putative protein product of Nbla00052; putative protein product of Nbla10993; TBC/LysM-associated domain containing 4
Gene Aliases: 9030406N13Rik; dJ187J11.3; ERAP140; ESNA1; Nbla00052; Nbla10993; NCOA7; NCOA7-AS; TLDC4
UniProt ID: (Human) Q8NI08, (Mouse) Q6DFV7
Entrez Gene ID: (Human) 135112, (Mouse) 211329
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support