Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SLC8A1 Antibodies

          Bioss

          NCX1/SLC8A1 Polyclonal Antibody, Alexa Fluor™ 350

          View all (21) SLC8A1 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite NCX1/SLC8A1 Polyclonal Antibody, Alexa Fluor™ 350

          Product Details

          BS-1550R-A350

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300-5,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:50-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          Flow Cytometry (Flow)

          1:20-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human NCX1
          View immunogen

          Conjugate

          Alexa Fluor™ 350 Alexa Fluor™ 350 Alexa Fluor™ 350

          Excitation/Emission Max

          343/441 nm View spectra spectra

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.03% ProClin 300

          Storage conditions

          -20°C or -80°C if preferred

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          Ca2+ plays a critical role in intracellular signaling and thus regulation of calcium balance is critical for the maintenance of this signaling function. The plasma membrane Na+/Ca2+ (NaCX) exchangers mediate Na+-dependent Ca2+ efflux in a wide variety of cell types. NaCX can move Ca2+ either into or out of cells, depending on the net Na+, Ca2+, and K+ gradient across the membrane. In mammals, at least 5 distinct genes code for the exchangers: Three NaCX (NaCX-1, NaCX-2, and NaCX-3), and two in the NaCKX family (NaCKX-1 and NaCKX-2). NaCX-1 is most prominently expressed in the heart where it plays a major role in excitation-contraction coupling (Matsuoka et al., 1993). NACX-1 may also be a target for protection of the heart and brain from ischemia and reperfusion injury (Hagihara et al., 2005; Bano et al., 2005; Choi, 2005).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Na(+)/Ca(2+)-exchange protein 1; Na+/Ca++ exchanger; Na+/Ca2+ exchanger; Na-Ca exchanger; Na/Ca exchanger; Sodium/calcium exchanger 1; sodium/calcium exchanger isoform NaCa7; solute carrier family 8 (sodium/calcium exchanger), member 1; Solute carrier family 8 member 1; solute carrier family 8, member 1

          View more View less

          Gene Aliases: AI852629; AV344025; CNC; D930008O12Rik; Ncx; NCX1; SLC8A1

          View more View less

          UniProt ID: (Human) P32418, (Mouse) P70414, (Rat) Q01728

          View more View less

          Entrez Gene ID: (Human) 6546, (Mouse) 20541, (Rat) 29715

          View more View less

          Function(s)
          calcium:sodium antiporter activity calcium ion binding protein binding calmodulin binding cytoskeletal protein binding ankyrin binding ion channel binding antiporter activity metal ion binding calcium:sodium antiporter activity involved in regulation of cardiac muscle cell membrane potential
          Process(es)
          response to hypoxia embryonic placenta development regulation of the force of heart contraction regulation of heart rate regulation of sodium ion transport heart morphogenesis transport ion transport sodium ion transport calcium ion transport cellular calcium ion homeostasis cellular sodium ion homeostasis cell communication positive regulation of cytosolic calcium ion concentration post-embryonic development positive regulation of fibroblast migration regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion vascular smooth muscle contraction positive regulation of bone mineralization response to ATP cellular response to reactive oxygen species embryonic heart tube development sodium ion transmembrane transport response to muscle stretch relaxation of smooth muscle muscle fiber development negative regulation of cytosolic calcium ion concentration regulation of calcium ion transport cardiac muscle cell development calcium ion homeostasis transmembrane transport cardiac muscle contraction calcium ion transport into cytosol calcium ion import calcium ion transmembrane transport cellular response to caffeine cellular response to cAMP sodium ion export regulation of cardiac muscle cell membrane potential cell communication by electrical coupling involved in cardiac conduction sodium ion import positive regulation of the force of heart contraction calcium ion export response to nutrient response to glucose telencephalon development response to immobilization stress response to drug response to hydrogen peroxide cellular response to hypoxia muscle contraction regulation of cell communication by electrical coupling regulation of cardiac muscle contraction by calcium ion signaling relaxation of cardiac muscle cytosolic calcium ion transport membrane depolarization during cardiac muscle cell action potential regulation of cardiac conduction
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67c77f6dc7-n6nf2:80/100.66.135.186:80.
          git-commit: 545e792480003146e539d624bd2a21b51d198228
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0