Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • NDE1 Antibodies

          Proteintech

          NDE1 Polyclonal Antibody, CoraLite® 594

          View all (15) NDE1 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite NDE1 Polyclonal Antibody, CoraLite® 594

          • Antibody Testing Data (2)
          NDE1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          NDE1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NDE1 Antibody (CL59410233100UL) in IHC (P)

          Immunofluorescent analysis of (4% PFA) fixed human breast cancer tissue using CoraLite 594 NDE1 antibody (Product # CL59410233100UL)) at dilution of 1:200. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NDE1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NDE1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Product Details

          CL59410233100UL

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Conjugate

          CoraLite® 594 CoraLite® 594 CoraLite® 594

          Excitation/Emission Max

          593/614 nm

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 0.5% BSA, 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles, store in dark

          Shipping conditions

          Wet ice

          Target Information

          NDE1 is s a member of the nuclear distribution E (NudE) family of proteins. It is localized at the centrosome and interacts with other centrosome components as part of a multiprotein complex that regulates dynein function. NDE1 protein plays an essential role in microtubule organization, mitosis and neuronal migration. Mutations to the gene lead to Lissencephaly 4 (LIS4) and Microhydranencephaly (MHAC).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: LIS1-interacting protein NUDE1; LIS1-interacting protein NUDE1, rat homolog; nuclear distribution gene E homolog 1; Nuclear distribution protein nudE homolog 1; NudE; nudE nuclear distribution E homolog 1; nudE nuclear distribution gene E homolog 1; rNudE

          View more View less

          Gene Aliases: 2810027M15Rik; AU042936; AW822251; HOM-TES-87; LIS4; MHAC; mNudE; NDE; NDE1; NUDE; NUDE1

          View more View less

          UniProt ID: (Human) Q9NXR1, (Rat) Q9ES39, (Mouse) Q9CZA6

          View more View less

          Entrez Gene ID: (Human) 54820, (Rat) 83836, (Mouse) 67203

          View more View less

          Function(s)
          protein binding microtubule binding protein domain specific binding identical protein binding
          Process(es)
          establishment of mitotic spindle orientation neuron migration microtubule nucleation chromosome segregation mitotic centrosome separation neuroblast proliferation cell migration cerebral cortex development forebrain development microtubule organizing center organization vesicle transport along microtubule centrosome duplication cell division establishment of chromosome localization centrosome localization cell cycle mitotic nuclear division multicellular organism development nervous system development cell differentiation G2/M transition of mitotic cell cycle sister chromatid cohesion regulation of microtubule motor activity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0