Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
This gene encodes a thiol-activated peptidase that is phosphorylated in M phase of the cell cycle. Phosphorylation regulates the cell cycle-dependent distribution of this protein, with a fraction of the protein bound strongly to centrosomes in interphase and localized to mitotic spindles in early M phase. Overall, this protein plays a role in nervous system development. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp451M0318; endooligopeptidase A; LIS1-interacting protein NUDEL; LIS1-interacting protein NUDEL; endooligopeptidase A; Mitosin-associated protein 1; mitosin-associated protein MITAP1; mNudE-L; nuclear disribution gene E like; nuclear distribution gene E-like homolog 1; Nuclear distribution protein nudE-like 1; nudE neurodevelopment protein 1-like 1; nudE nuclear distribution E homolog (A. nidulans)-like 1; nudE nuclear distribution E homolog-like 1; nudE nuclear distribution gene E homolog (A. nidulans)-like 1; nudE nuclear distribution gene E homolog like 1; nudE nuclear distribution gene E homolog-like 1; NUDE-like protein; Protein mNudE-like; Protein Nudel
Gene Aliases: 2600006O07Rik; EOPA; MITAP1; mNudel; NDE1L1; NDE2; NDEL1; NUDE2; NUDEL
UniProt ID: (Human) Q9GZM8, (Rat) Q78PB6, (Mouse) Q9ERR1
Entrez Gene ID: (Human) 81565, (Dog) 489495, (Rat) 170845, (Mouse) 83431
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support