Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAELQEVQI TEEKPLLPGQ TPEAAKTHSV ETPYVSVTFT VYGTPKPKRP AILTYHDVGL NYKSCFQPLF QFEDMQEIIQ NFVRVHVDAP GMEEGAPVFP LGYQYPSLDQ LADMIPCVLQ YLNFSTIIGV GVGAGAYILA RYALNHPDTV EGLVLINIDP NAKGWMDWAA HKLTGLTSSI PEMILGHLFS QEELSGNSEL IQKYRNIITH APNLDNIELY WNSYNNRRDL NFERGGDITL RCPVMLVVGD QAPHEDAVVE CNSKLDPTQT SFLKMADSGG QPQLTQPGKL TEAFKYFLQG MGYMASSCMT RLSRSRTASL TSAASVDGNR SRSRTLSQSS ESGTLSSGPP GHTMEVSC (1-357 aa encoded by BC010458)
NDRG2's gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. NDRG2 is a cytoplasmic protein that may play a role in neurite outgrowth. Its gene may be involved in glioblastoma carcinogenesis.This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may play a role in neurite outgrowth. This gene may be involved in glioblastoma carcinogenesis. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antidepressant-related protein ADRG123; cytoplasmic protein Ndr1; DKFZp781G1938; FLJ25522; N-myc downstream regulated 2; N-myc downstream regulated gene 2; N-myc downstream regulator 2; N-myc downstream-regulated gene 2 protein; NDR1-related protein NDR2; NDRG1-related protein; NDRG2var; Protein Ndr2; Protein NDRG2; Protein Syld709613; syld709613 protein
Gene Aliases: AI182517; AU040374; KIAA1248; Ndr2; NDRG2; SYLD
UniProt ID: (Human) Q9UN36, (Mouse) Q9QYG0, (Rat) Q8VBU2
Entrez Gene ID: (Human) 57447, (Mouse) 29811, (Rat) 171114
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support