Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MELVQVLKRG LQQITGHGGL RGYLRVFFRT NDAKVGTLVG EDKYGNKYYE DNKQFFGRHR WVVYTTEMNG KNTFWDVDGS MVPPEWHRWL HSMTDDPPTT KPLTARKFIW TNHKFNVTGT PEQYVPYSTT RKKIQEWIPP STPYK; Positive Samples: 293T, PC-12, HepG2, Mouse kidney, Mouse heart, Mouse testis, Rat heart, Rat brain; Cellular Location: Matrix side, Mitochondrion inner membrane, Peripheral membrane protein
This gene encodes a protein which is part of mitochondrial complex 1, part of the oxidative phosphorylation system in mitochondria. Complex 1 transfers electrons to ubiquinone from NADH which establishes a proton gradient for the generation of ATP. Mutations in this gene are associated with Leigh syndrome due to mitochondrial complex 1 deficiency. Pseudogenes of this gene are located on chromosomes 5 and 13. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 13 kDa differentiation-associated protein; 13kDa differentiation-associated protein; CI-B17.2; CIB17.2; complex I B17.2 subunit; Complex I-B17.2; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 12; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 12; NADH-ubiquinone oxidoreductase subunit B17.2
Gene Aliases: 2410011G03Rik; AW112974; B17.2; DAP13; NDUFA12; RGD1311462
UniProt ID: (Human) Q9UI09, (Mouse) Q7TMF3
Entrez Gene ID: (Human) 55967, (Rat) 299739, (Mouse) 66414
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support