Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-13kD-B; complex I 13kDa subunit B; Complex I subunit B13; Complex I-13kD-B; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5, 13kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 5; NADH-ubiquinone oxidoreductase 13 kDa-B subunit; type I dehydrogenase; ubiquinone reductase
Gene Aliases: B13; CI-13kB; CI-13KD-B; NDUFA5; NUFM; UQOR13
UniProt ID: (Human) Q16718
Entrez Gene ID: (Human) 4698
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support