Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MASRVLSAYV SRLPAAFAPL PRVRMLAVAR PLSTALCSAG TQTRLGTLQP ALVLAQVPGR VTQLCRQYSD MPPLTLEGIQ DRVLYVLKLY DKIDPEKLSV NSHFMKDLGL DSLDQVEIIM AMEDEFGFEI PDIDAEKLMC PQEIVDYIAD KKDVYE
Carrier of the growing fatty acid chain in fatty acid biosynthesis in mitochondria. Accessory and non-catalytic subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), which functions in the transfer of electrons from NADH to the respiratory chain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACP; Acyl carrier protein, mitochondrial; CI-SDAP; complex I SDAP subunit; mitochondrial acyl carrier protein; NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1, 8kDa; NADH-ubiquinone oxidoreductase 9.6 kDa subunit; NADH:ubiquinone oxidoreductase SDAP subunit
Gene Aliases: ACP; FASN2A; NDUFAB1; SDAP
UniProt ID: (Human) O14561
Entrez Gene ID: (Human) 4706
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support