Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EIPEGYVPEH WEYYKHPISR WIARNFYDSP EKIYERTMAV LQIEAEKAEL RVKELEVRKL MHVRGDGPWY YYETIDKELI DHSPKATPDN; Positive Samples: HepG2, mouse kidney, rat kidney; Cellular Location: Matrix side, Mitochondrion inner membrane, Single-pass membrane protein
NDUFB5 is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. The protein encoded by this gene is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-SGDH; complex I SGDH subunit; Complex I-SGDH; DKFZp686N02262; FLJ30597; MGC111204; MGC12314; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 5, mitochondrial; NADH-ubiquinone oxidoreductase SGDH subunit
Gene Aliases: 0610007D05Rik; AU015782; CISGDH; NDUFB5; SGDH
UniProt ID: (Human) O43674, (Mouse) Q9CQH3
Entrez Gene ID: (Human) 4711, (Mouse) 66046, (Rat) 294964
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support