Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Sequence of this protein is as follows: MAALRALCGF RGVAAQVLRP GAGVRLPIQP SRGVRQWQPD VEWAQQFGGA VMYPSKETAH WKPPPWNDVD PPKDTIVKNI TLNFGPQHPA AHGVLRLVME LSGEMVRKCD PHIGLLHRGT EKLIEYKTYL QALPYFDRLD YVSMMCNEQA YSLAVEKLLN IRPPPRAQWI RVLFGEITRL LNHIMAVTTH ALDLGAMTPF FWLFEEREKM FEFYERVSGA RMHAAYIRPG GVHQDLPLGL MDDIYQFSKN FSLRLDELEE LLTNNRIWRN RTIDIGVVTA EEALNYGFSG VMLRGSGIQW DLRKTQPYDV YDQVEFDVPV GSRGDCYDRY LCRVEEMRQS LRIIAQCLNK MPPGEIKVDD AKVSPPKRAE MKTSMESLIH HFKLYTEGYQ VPPGATYTAI EAPKGEFGVY LVSDGSSRPY RCKIKAPGFA HLAGLDKMSK GHMLADVVAI IGTQDIVFGE VDR
The protein encoded by this gene is a core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase. Mammalian mitochondrial complex I is composed of at least 43 different subunits, 7 of which are encoded by the mitochondrial genome, and the rest are the products of nuclear genes. The iron-sulfur protein fraction of complex I is made up of 7 subunits, including this gene product. Complex I catalyzes the NADH oxidation with concomitant ubiquinone reduction and proton ejection out of the mitochondria. Mutations in this gene are associated with mitochondrial complex I deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-49kD; comp; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 49kDa subunit; Complex I-49kD; NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial; NADH-ubiquinone oxidoreductase 49 kDa subunit; NADH-ubiquinone oxidoreductase NDUFS2 subunit
Gene Aliases: CI-49; NDUFS2
UniProt ID: (Human) O75306
Entrez Gene ID: (Human) 4720
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support