Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MAAAAVARLW WRGILGASAL TRGTGRPSVL LLPVRRESAG ADTRPTVRPR NDVAHKQLSA FGEYVAEILP KYVQQVQVSC FNELEVCIHP DGVIPVLTFL RDHTNAQFKS LVDLTAVDVP TRQNRFEIVY NLLSLRFNSR IRVKTYTDEL TPIESAVSVF KAANWYEREI WDMFGVFFAN HPDLRRILTD YGFEGHPFRK DFPLSGYVEL RYDDEVKRVV AEPVELAQEF RKFDLNSPWE AFPVYRQPPE SLKLEAGDKK PDAK
This gene encodes one of the iron-sulfur protein components of mitochondrial NADH:ubiquinone oxidoreductase. Mutations in this gene are associated with Leigh syndrome resulting from mitochondrial complex I deficiency.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-30kD; complex I 30kDa subunit; Complex I-30kD; NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial; NADH dehydrogenase-ubiquinone 30 kDa subunit; NADH-ubiquinone oxidoreductase 30 kDa subunit
Gene Aliases: CI-30; NDUFS3
UniProt ID: (Human) O75489
Entrez Gene ID: (Human) 4722
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support