Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MLATRRLLGW SLPARVSVRF SGDTTAPKKT SFGSLKDEDR IFTNLYGRHD WRLKGSLSRG DWYKTKEILL KGPDWILGEI KTSGLRGRGG AGFPTGLKWS FMNKPSDGRP KYLVVNADEG EPGTCKDREI LRHDPHKLLE GCLVGGRAMG ARAAYIYIRG EFYNEASNLQ VAIREAYEAG LIGKNACGSG YDFDVFVVRG AGAYICGEET ALIESIEGKQ GKPRLKPPFP ADVGVFGCPT TVANVETVAV SPTICRRGGT WFAGFGRERN SGTKLFNISG HVNHPCTVEE EMSVPLKELI EKHAGGVTGG WDNLLAVIPG GSSTPLIPKS VCETVLMDFD ALVQAQTGLG TAAVIVMDRS TDIVKAIARL IEFYKHESCG QCTPCREGVD WMNKVMARFV RGDARPAEID SLWEISKQIE GHTICALGDG AAWPVQGLIR HFRPELEERM QRFAQQHQAR QAAS
NDUFV1 is the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.The NDUFV1 gene encodes the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CI-51kD; complex I 51 kda subunit; complex I 51kDa subunit; complex I, mitochondrial respiratory chain; Complex I-51kD; mitochondrial NADH dehydrogenase ubiquinone flavoprotein 1; mitochondrial NADH:ubiquinone oxidoreductase 51 kda subunit; NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa; NADH dehydrogenase [ubiquinone] flavoprotein 1, mitochondrial; NADH dehydrogenase flavoprotein 1; NADH-ubiquinone oxidoreductase 51 kDa subunit; NDUFV1
Gene Aliases: CI-51K; CI51KD; NDUFV1; UQOR1
UniProt ID: (Human) P49821
Entrez Gene ID: (Human) 4723
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support