Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MATELEYESV LCVKPDVSVY RIPPRASNRG YRASDWKLDQ PDWTGRLRIT SKGKTAYIKL EDKVSGELFA QAPVEQYPGI AVETVTDSSR YFVIRIQDGT GRSAFIGIGF TDRGDAFDFN VSLQDHFKWV KQESEISKES QEMDARPKLD LGFKEGQTIK LCIGNITNKK GGASKPRTAR GGGLSLLPPP PGGKVTIPPP SSSVAISNHV TPPPIPKSNH GGSDADILLD LDSPAPVTTP APTPVSVSND LWGDFSTASS SVPNQAPQPS NWVQF
Involved in endocytosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Adaptin ear-binding coat-associated protein 1; NECAP endocytosis-associated protein 1; NECAP-1
Gene Aliases: EIEE21; NECAP1
UniProt ID: (Human) Q8NC96
Entrez Gene ID: (Human) 25977
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support