Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
In direct ELISAs, approximately 25% cross-reactivity with recombinant human NELL1 is observed and approximately 10% cross-reactivity with recombinant mouse NELL2 is observed.
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
This gene encodes a cytoplasmic protein that contains epidermal growth factor (EGF)-like repeats. The encoded heterotrimeric protein may be involved in cell growth regulation and differentiation. A similar protein in rodents is involved in craniosynostosis. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NEL-like protein 1; Protein kinase C-binding protein NELL1
Gene Aliases: B230343H07Rik; l7R6; Nell1
UniProt ID: (Mouse) Q2VWQ2
Entrez Gene ID: (Mouse) 338352
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support