Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAAASEPVD SGALWGLERP EPPPTRFHRV HGANIRVDPS GTRATRVESF AHGVCFSREP LAPGQVFLVE IEEKELGWCG HLRLGLTALD PASLAPVPEF SLPDLVNLGH TWVFAITRHH NRVPREGRPE AEAAAPSRPP TLLVEPYLRI EQFRIPRDRL VGRSRPGLYS HLLDQLYELN VLPPTARRSR LGVLFCPRPD GTADMHIIIN GEDMGPSARG LPAAQPLYAV VDVFASTKSV RLVQLEYGLP SLQTLCRLVI QRSMVHRLAI DGLHLPKELK DFCKYE (1-285 aa encoded by B C105935)
This gene encodes a protein that is involved in the regulation of myofibril organization. This protein is likely the adaptor component of the E3 ubiquitin ligase complex in striated muscle, and it regulates the ubiquitin-mediated degradation of beta-catenin during myogenesis. Alternative splicing of this gene results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: neuralized homolog 2; neuralized-like 2; Neuralized-like protein 2
Gene Aliases: C20orf163; Neur2; NEURL2; OZZ; OZZ-E3
UniProt ID: (Human) Q9BR09
Entrez Gene ID: (Human) 140825, (Mouse) 415115, (Rat) 311633
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support