Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: QDMPPHLPTA SASFPVHPYS YQSPGLPSPP YGTMDSSHVF HVKPPPHAYS AALEPFFESP LTDCTSPSFD GPLSPPLSIN GNFSFKHEPS AEFEKNYAFT
NeuroD1 is a transcriptional activator that acts as a differentiation factor during neurogenesis. It has been demonstrated to bind to the insulin gene E-box. Efficient DNA binding requires dimerization with another basic helix-loop-helix (bHLH) protein. Defects in NEUROD1 are a cause of maturity onset diabetes of the young type VI (MODY6). MODY6 is a form of non-insulin-dependent diabetes mellitus characterized by an autosomal dominant mode of inheritance, onset during young adulthood and a primary defect in insulin secretion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: basic helix-loop-helix transcription factor; beta-cell E-box transactivator 2; BHF1; bHLHa3; Class A basic helix-loop-helix protein 3; neuro d1; NeuroD; Neurogenic differentiation factor 1; neurogenic helix-loop-helix protein NEUROD
Gene Aliases: BETA2; BHF-1; BHLHA3; MODY6; NEUROD; NEUROD1
UniProt ID: (Human) Q13562
Entrez Gene ID: (Human) 4760
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support