Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: GQGGEAAHHT RSPYSTFYPP YHSPELTTPP GHGTLDNSKS MKPYNYCSAY ESFYESTSPE CASPQFEGPL SPPPINYNGI FSLKQEETLD YGKNYNYGMH
NEUROD6 contains 1 basic helix-loop-helix (bHLH) domain. It activates E box-dependent transcription in collaboration with TCF3/E47 and may be a trans-acting factor involved in the development and maintenance of the mammalian nervous system. It transactivates the promoter of its own gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHa2; brain my051 protein; Class A basic helix-loop-helix protein 2; NeuroD6; Neurogenic differentiation factor 6; Protein atonal homolog 2
Gene Aliases: ATOH2; BHLHA2; Math-2; MATH2; My051; NEUROD6; Nex1; NEX1M
UniProt ID: (Human) Q96NK8
Entrez Gene ID: (Human) 63974
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support